Boc-DL-2-aminoheptanedioic acid

Supplier: Matrix Scientific

041636-1G 041636-5G
101852-574EA 412.31 USD
101852-574 101852-576
Boc-DL-2-aminoheptanedioic acid
Boc-DL-2-aminoheptanedioic acid

Hazard: Irritant

Formula: C₁₂H₂₁NO₆
MDL Number: MFCD09750509
CAS Number: 1027776-55-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR