Boc-D-2-aminoadipic acid

Supplier: Matrix Scientific

041634-1G 041634-5G
101852-566EA 498.46 USD
101852-566 101852-568
Boc-D-2-aminoadipic acid
Boc-D-2-aminoadipic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₆
MDL Number: MFCD00797554
CAS Number: 110544-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR