Boc-α-methyl-L-aspartic acid

Supplier: Matrix Scientific

041632-100MG 041632-500MG
101852-558EA 536.92 USD
101852-558 101852-560
Boc-α-methyl-L-aspartic acid
Boc-alpha-methyl-L-aspartic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₆
MW: 247.25 g/mol
MDL Number: MFCD02682501
CAS Number: 1217679-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR