Boc-α-methyl-DL-glutamic acid

Supplier: Matrix Scientific

041631-1G 041631-5G
101852-554EA 735.38 USD
101852-554 101852-556
Boc-α-methyl-DL-glutamic acid
Boc-alpha-methyl-DL-glutamic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₆
MW: 261.28 g/mol
MDL Number: MFCD02682511
CAS Number: 189149-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR