Boc-2,6-diaminopimelic acid

Supplier: Matrix Scientific

041629-1G 041629-5G
101852-546EA 386.15 USD
101852-546 101852-548
Boc-2,6-diaminopimelic acid
Boc-2,6-diaminopimelic acid

Hazard: Irritant

Formula: C₁₇H₃₀N₂O₈
MDL Number: MFCD09750513
CAS Number: 98469-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR