Boc-[N-α-(4-aminobenzoyl)]-L-glutamic acid
Supplier: Matrix Scientific
041628-1G
041628-5G
101852-542EA
386.15
USD
101852-542
101852-544
Boc-[N-α-(4-aminobenzoyl)]-L-glutamic acid
Boc-[N-alpha-(4-aminobenzoyl)]-L-glutamic acid
Hazard: Irritant
Formula:
C₁₇H₂₂N₂O₇ MW: 366.37 g/mol |
MDL Number:
MFCD04974554 CAS Number: 1562425-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...