Boc-(N-γ-ethyl)-L-glutamine

Supplier: Matrix Scientific

041627-1G 041627-5G
101852-538EA 686.15 USD
101852-538 101852-540
Boc-(N-γ-ethyl)-L-glutamine
Boc-(N-gamma-ethyl)-L-glutamine

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₅
MW: 274.32 g/mol
MDL Number: MFCD00153304
CAS Number: 210692-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR