Fmoc-Arg(NO2)-OH

Supplier: Matrix Scientific

Synonyms: Nα-Fmoc-Nomega-nitro-L-arginine, Fmoc-Arg(NO₂)-OH

041626-1G 041626-5G
101852-534EA 87.69 USD
101852-534 101852-536
Fmoc-Arg(NO2)-OH
Fmoc-Arg(NO2)-OH

Hazard: Irritant

Formula: C₂₁H₂₃N₅O₆
MDL Number: MFCD00065617
CAS Number: 58111-94-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR