Fmoc-Nw-nitro-D-arginine

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Arg(NO2)-OH, Fmoc-D-Arg(NO₂)-OH, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(3-nitroguanidino)pentanoic acid

041625-1G 041625-5G
101852-530EA 412.31 USD
101852-530 101852-532
Fmoc-Nw-nitro-D-arginine
Fmoc-Nw-nitro-D-arginine

Hazard: Irritant

Formula: C₂₁H₂₃N₅O₆
MW: 441.45 g/mol
MDL Number: MFCD02682438
CAS Number: 160347-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR