(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid

Supplier: Matrix Scientific

Synonyms: Fmoc-Arg(Me)₂-OH (symmetrical), Fmoc-Arg(Me, Me)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((bis(methylamino)methylene)amino)pentanoic acid

041624-250MG 041624-1G
101852-526EA 498.46 USD
101852-526 101852-528
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid
(S,E)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(2,3-dimethylguanidino)pentanoic acid

Hazard: Irritant

Formula: C₂₃H₂₈N₄O₄
MW: 424.501 g/mol
MDL Number: MFCD02684483
CAS Number: 823780-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR