Fmoc-Arg(Me)2-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-Arg(Me)₂-OH (asymmetrical)
041623-250MG
041623-1G
101852-522EA
326.15
USD
101852-522
101852-524
Fmoc-Arg(Me)2-OH
Fmoc-Arg(Me)2-OH
Hazard: Irritant
Formula:
C₂₃H₂₈N₄O₄ MW: 424.501 g/mol |
MDL Number:
MFCD02684510 CAS Number: 268564-10-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...