Fmoc-Lys(Me3)-OH chloride

Supplier: Matrix Scientific

Synonyms: Fmoc-Lys(Me)₃-OH chloride, Fmoc-Lys(Me)3-OH Chloride, Fmoc-Lys(Me)3-OH Chloride

041622-250MG 041622-500MG 041622-1G
101852-516EA 661.54 USD
101852-516 101852-518 101852-520
Fmoc-Lys(Me3)-OH chloride
Fmoc-Lys(Me3)-OH chloride

Hazard: Irritant

Formula: C₂₄H₃₁ClN₂O₄
MW: 446.972 g/mol
MDL Number: MFCD00672333
CAS Number: 201004-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR