Fmoc-Lys(Me)₂-OH·HCl

Supplier: Matrix Scientific

Synonyms: N-α-Fmoc-N-ε-dimethyl-N-L-lysine hydrochloride, Fmoc-Lys(Me2)-OH.HCl

041621-250MG 041621-1G
101852-512EA 312.31 USD
101852-512 101852-514
Fmoc-Lys(Me)₂-OH·HCl
Fmoc-Lys(ME)2-OH·HCl

Hazard: Irritant

Formula: C₂₃H₂₉ClN₂O₄
MW: 432.945 g/mol
MDL Number: MFCD05662353
CAS Number: 252049-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR