(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid

Supplier: Matrix Scientific

Synonyms: N-Fmoc-L-citrulline, Nalpha-Fmoc-L-citrulline, Fmoc-Cit-OH

041620-1G 041620-5G
101852-508EA 15.38 USD
101852-508 101852-510
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid

Hazard: Irritant

Formula: C₂₁H₂₃N₃O₅
MW: 397.4 g/mol
MDL Number: MFCD00151943
CAS Number: 133174-15-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR