Fmoc-L-2-amino-3-guanidinopropionic acid

Supplier: Matrix Scientific

041618-250MG 041618-1G
101852-500EA 786.15 USD
101852-500 101852-502
Fmoc-L-2-amino-3-guanidinopropionic acid
Fmoc-L-2-amino-3-guanidinopropionic acid

Hazard: Irritant

Formula: C₁₉H₂₀N₄O₄
MDL Number: MFCD02682426
CAS Number: 1217839-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR