Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine

Supplier: Matrix Scientific

Synonyms: Nδ-Boc-Nɑ-Fmoc-L-ornithine, Nδ-Boc-Nα-Fmoc-L-ornithine, Fmoc-Orn(Boc)-OH

041616-5G 041616-25G
101852-492EA 52.31 USD
101852-492 101852-494
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine
Ndelta-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine

Hazard: Irritant

Formula: C₂₅H₃₀N₂O₆
MW: 454.52 g/mol
MDL Number: MFCD00065668
CAS Number: 109425-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR