Fmoc-D-Orn(Boc)-OH

Supplier: Matrix Scientific

Synonyms: Nδ-Boc-Nα-Fmoc-D-ornithine

041615-1G 041615-5G 041615-25G
101852-486EA 150.77 USD
101852-486 101852-488 101852-490
Fmoc-D-Orn(Boc)-OH
Fmoc-D-Orn(Boc)-OH

Hazard: Irritant

Formula: C₂₅H₃₀N₂O₆
MDL Number: MFCD00077065
CAS Number: 118476-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR