Fmoc-Orn(Dde)-OH

Supplier: Matrix Scientific

041612-1G 041612-5G
101852-474EA 461.54 USD
101852-474 101852-476
Fmoc-Orn(Dde)-OH
Fmoc-Orn(Dde)-OH

Hazard: Irritant

Formula: C₃₀H₃₄N₂O₆
MDL Number: MFCD01862895
CAS Number: 269062-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR