Fmoc-(Nd-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-ornithine

Supplier: Matrix Scientific

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-((1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethyl)amino)pentanoic acid, Fmoc-D-Orn(Dde)-OH

041611-1G 041611-5G
101852-470EA 498.46 USD
101852-470 101852-472
Fmoc-(Nd-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-ornithine
Fmoc-(Nd-1-(4,4-dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl)-D-ornithine

Hazard: Irritant

Formula: C₃₀H₃₄N₂O₆
MW: 518.62 g/mol
MDL Number: MFCD02094127
CAS Number: 1419640-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR