Di-fmoc-α-methyl-DL-ornithine

Supplier: Matrix Scientific

041609-100MG 041609-500MG
101852-462EA 823.08 USD
101852-462 101852-464
Di-fmoc-α-methyl-DL-ornithine
Di-fmoc-alpha-methyl-DL-ornithine

Hazard: Irritant

Formula: C₃₆H₃₄N₂O₆
MW: 590.68 g/mol
MDL Number: MFCD04974550
CAS Number: 1219207-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR