Di-fmoc-2,6-diaminoheptanedioic acid (mixture ofisomers)

Supplier: Matrix Scientific

041608-1G 041608-5G
101852-458EA 498.46 USD
101852-458 101852-460
Di-fmoc-2,6-diaminoheptanedioic acid (mixture ofisomers)
Di-fmoc-2,6-diaminoheptanedioic acid (mixture ofisomers)

Hazard: Irritant

Formula: C₃₇H₃₄N₂O₈
MDL Number: MFCD02682434
CAS Number: 1198797-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR