Di-boc-α-methyl-DL-ornithine

Supplier: Matrix Scientific

041607-100MG 041607-500MG
101852-454EA 823.08 USD
101852-454 101852-456
Di-boc-α-methyl-DL-ornithine
Di-boc-alpha-methyl-DL-ornithine

Hazard: Irritant

Formula: C₁₆H₃₀N₂O₆
MW: 346.43 g/mol
MDL Number: MFCD04974549
CAS Number: 1219378-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR