Di-boc-2,6-diaminoheptanedioic acid (mixture of isomers)

Supplier: Matrix Scientific

041606-1G 041606-5G
101852-450EA 498.46 USD
101852-450 101852-452
Di-boc-2,6-diaminoheptanedioic acid (mixture of isomers)
Di-boc-2,6-diaminoheptanedioic acid (mixture of isomers)

Hazard: Irritant

Formula: C₁₇H₃₀N₂O₈
MDL Number: MFCD02682433
CAS Number: 98469-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR