Boc-D-Arg(NO₂)-OH ≥95%

Supplier: Matrix Scientific

Synonyms: N2-(tert-Butoxycarbonyl)-Nw-nitro-D-arginine

041604-1G 041604-5G
101852-442EA 80 USD
101852-442 101852-444
Boc-D-Arg(NO₂)-OH ≥95%
Boc-D-Arg(NO₂)-OH

Hazard: Irritant

Formula: C₁₁H₂₁N₅O₆
MDL Number: MFCD00038091
CAS Number: 50913-12-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR