Boc-Lys(N3)-OH

Supplier: Matrix Scientific

041601-250MG 041601-1G
101852-430EA 413.85 USD
101852-430 101852-432
Boc-Lys(N3)-OH
Boc-Lys(N3)-OH

Hazard: Irritant

Formula: C₁₁H₂₀N₄O₄
MW: 272.31 g/mol
MDL Number: MFCD09750502
CAS Number: 846549-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR