Boc-Cit-OH

Supplier: Matrix Scientific

Synonyms: (S)-2-((tert-Butoxycarbonyl)amino)-5-ureidopentanoic acid

041600-1G 041600-5G
101852-426EA 130.77 USD
101852-426 101852-428
Boc-Cit-OH
Boc-Cit-OH

Hazard: Irritant

Formula: C₁₁H₂₁N₃O₅
MW: 275.305 g/mol
MDL Number: MFCD00080271
CAS Number: 45234-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR