Boc-L-2-amino-3-ureidopropionic acid

Supplier: Matrix Scientific

041599-250MG 041599-1G
101852-422EA 275.38 USD
101852-422 101852-424
Boc-L-2-amino-3-ureidopropionic acid
Boc-L-2-amino-3-ureidopropionic acid

Hazard: Irritant

Formula: C₉H₁₇N₃O₅
MDL Number: MFCD02682427
CAS Number: 901284-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR