Boc-L-2-amino-3-guanidinopropionic acid

Supplier: Matrix Scientific

041598-250MG 041598-1G
101852-418EA 786.15 USD
101852-418 101852-420
Boc-L-2-amino-3-guanidinopropionic acid
Boc-L-2-amino-3-guanidinopropionic acid

Hazard: Irritant

Formula: C₉H₁₈N₄O₄
MDL Number: MFCD02682425
CAS Number: 1217718-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR