Fmoc-L-norvaline

Supplier: Matrix Scientific

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, Fmoc-Nva-OH, N-Fmoc-L-norvaline

041596-5G 041596-25G
101852-410EA 104.62 USD
101852-410 101852-412
Fmoc-L-norvaline
Fmoc-L-norvaline

Hazard: Irritant

Formula: C₂₀H₂₁NO₄
MW: 339.391 g/mol
MDL Number: MFCD00155631
CAS Number: 135112-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR