Fmoc-N(Hmb)-Gly-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-(Hmb)Gly-OH

041595-1G 041595-5G
101852-406EA 516.92 USD
101852-406 101852-408
Fmoc-N(Hmb)-Gly-OH
Fmoc-N(Hmb)-Gly-OH

Hazard: Irritant

Formula: C₂₅H₂₃NO₆
MW: 433.46 g/mol
MDL Number: MFCD03093500
CAS Number: 148515-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR