Fmoc-L-propargylglycine ≥95%

Supplier: Matrix Scientific

041594-1G 041594-5G
101852-402EA 90.77 USD
101852-402 101852-404
Fmoc-L-propargylglycine ≥95%
Fmoc-L-propargylglycine
Formula: C₂₀H₁₇NO₄
MDL Number: MFCD01075095
CAS Number: 198561-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR