N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylacetic acid, N-Fmoc-L-α-phenylglycine, Fmoc-Phg-OH, N-Fmoc-L-2-phenylglycine

041593-5G 041593-25G
101852-400EA 18.46 USD
101852-400 102663-704
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MW: 373.41 g/mol
MDL Number: MFCD00155632
CAS Number: 102410-65-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR