Fmoc-L-homocyclohexylalanine
Supplier: Matrix Scientific
Synonyms:
Fmoc-L-Homocyclohexylalanine
041592-1G
041592-5G
101852-394EA
412.31
USD
101852-394
101852-396
Fmoc-L-homocyclohexylalanine
Fmoc-L-homocyclohexylalanine
Hazard: Irritant
Formula:
C₂₅H₂₉NO₄ |
MDL Number:
MFCD02179645 CAS Number: 269078-73-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...