Fmoc-Chg-OH

Supplier: Matrix Scientific

Synonyms: Fmoc-L-Chg-OH, Fmoc-cyclohexyl-Gly-OH, N-Fmoc-L-cyclohexylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-cyclohexylglycine, N-Fmoc-2-cyclohexyl-L-glycine

041591-1G 041591-5G
101852-390EA 106.15 USD
101852-390 101852-392
Fmoc-Chg-OH
Fmoc-Chg-OH

Hazard: Irritant

Formula: C₂₃H₂₅NO₄
MW: 379.456 g/mol
MDL Number: MFCD00190878
CAS Number: 161321-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR