Fmoc-Tle-OH

Supplier: Matrix Scientific

041590-1G 041590-5G
101852-386EA 83.08 USD
101852-386 101852-388
Fmoc-Tle-OH
Fmoc-Tle-OH

Hazard: Irritant

Formula: C₂₁H₂₃NO₄
MDL Number: MFCD00065649
CAS Number: 132684-60-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR