Fmoc-Dab-OH
Supplier: Matrix Scientific
Synonyms:
(S)-2-(Fmoc-amino)-4-aminobutyric acid
041588-1G
041588-5G
101852-378EA
187.69
USD
101852-378
101852-380
Fmoc-Dab-OH
Fmoc-Dab-OH
Hazard: Irritant
Formula:
C₁₉H₂₀N₂O₄ MW: 340.379 g/mol |
MDL Number:
MFCD00237017 CAS Number: 161420-87-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...