Fmoc-Dap-OH

Supplier: Matrix Scientific

041587-1G 041587-5G
101852-374EA 52.31 USD
101852-374 101852-376
Fmoc-Dap-OH
Fmoc-Dap-OH

Hazard: Irritant

Formula: C₁₈H₁₈N₂O₄
MDL Number: MFCD00237019
CAS Number: 181954-34-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR