Fmoc-L-allylglycine

Supplier: Matrix Scientific

Synonyms: Fmoc-Gly(allyl)-OH

041586-1G 041586-5G
101852-370EA 498.46 USD
101852-370 101852-372
Fmoc-L-allylglycine
Fmoc-L-allylglycine

Hazard: Irritant

Formula: C₂₀H₁₉NO₄
MW: 337.38 g/mol
MDL Number: MFCD01311749
CAS Number: 146549-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR