Fmoc-L-2-indanylglycine

Supplier: Matrix Scientific

Synonyms: (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(2,3-dihydro-1H-inden-2-yl)acetic acid

041585-1G 041585-5G
101852-366EA 498.46 USD
101852-366 101852-368
Fmoc-L-2-indanylglycine
Fmoc-L-2-indanylglycine

Hazard: Irritant

Formula: C₂₆H₂₃NO₄
MDL Number: MFCD00672561
CAS Number: 205526-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR