Fmoc-L-2-amino-3-(dimethylamino)propionic acid

Supplier: Matrix Scientific

041583-500MG 041583-1G
101852-358EA 2230.77 USD
101852-358 101852-360
Fmoc-L-2-amino-3-(dimethylamino)propionic acid
Formula: C₂₀H₂₂N₂O₄
MDL Number: MFCD02682424
CAS Number: 587880-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR