Fmoc-L-1-pyrenylalanine

Supplier: Matrix Scientific

041582-250MG 041582-1G
101852-354EA 536.92 USD
101852-354 101852-356
Fmoc-L-1-pyrenylalanine
Fmoc-L-1-pyrenylalanine

Hazard: Irritant

Formula: C₃₄H₂₅NO₄
MW: 511.58 g/mol
MDL Number: MFCD01861376
CAS Number: 183071-07-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR