Fmoc-L-(3-thienyl)glycine

Supplier: Matrix Scientific

041581-100MG 041581-500MG
101852-350EA 598.46 USD
101852-350 101852-352
Fmoc-L-(3-thienyl)glycine
Fmoc-L-(3-thienyl)glycine

Hazard: Irritant

Formula: C₂₁H₁₇NO₄S
MDL Number: MFCD02682480
CAS Number: 1217706-09-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR