Fmoc-GABA-OH

Supplier: Matrix Scientific

Synonyms: 4-(Fmoc-amino)butyric acid, N-Fmoc-4-aminobutyric acid, Fmoc-γ-Abu-OH, Fmoc-GABA-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-aminobutyric acid

041580-1G 041580-5G
101852-346EA 113.85 USD
101852-346 101852-348
Fmoc-GABA-OH
Fmoc-GABA-OH

Hazard: Irritant

Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
MDL Number: MFCD00144889
CAS Number: 116821-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR