Fmoc-D-Phg-OH ≥95%

Supplier: Matrix Scientific

Synonyms: Fmoc-D-Phg-OH, N-Fmoc-D-α-phenylglycine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-2-phenylglycine, N-Fmoc-D-2-phenylglycine

041578-5G 041578-25G
101852-340EA 69.23 USD
101852-340 102551-290
Fmoc-D-Phg-OH ≥95%
Fmoc-D-Phg-OH

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MW: 373.408 g/mol
MDL Number: MFCD00190891
CAS Number: 111524-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR