Fmoc-D-Nva-OH
Supplier: Matrix Scientific
041577-5G
041577-25G
101852-334EA
224.62
USD
101852-334
101852-336
Fmoc-D-Nva-OH
Fmoc-D-Nva-OH
Hazard: Irritant
Formula:
C₂₀H₂₁NO₄ |
MDL Number:
MFCD00155640 CAS Number: 144701-24-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...