Fmoc-DL-β-aminoisobutyric acid
Supplier: Matrix Scientific
041576-1G
041576-5G
101852-330EA
326.15
USD
101852-330
101852-332
Fmoc-DL-β-aminoisobutyric acid
Fmoc-DL-beta-aminoisobutyric acid
Hazard: Irritant
Formula:
C₁₉H₁₉NO₄ MW: 325.37 g/mol |
MDL Number:
MFCD01863213 CAS Number: 186320-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...