Fmoc-DL-β-aminobutyric acid
Supplier: Matrix Scientific
Synonyms:
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)butanoic acid
041575-1G
041575-5G
101852-326EA
380
USD
101852-326
101852-328
Fmoc-DL-β-aminobutyric acid
Fmoc-DL-beta-aminobutyric acid
Hazard: Irritant
Formula:
C₁₉H₁₉NO₄ MW: 325.37 g/mol |
MDL Number:
MFCD00270340 CAS Number: 186320-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...