Fmoc-DL-2-amino-3-(dimethylamino)propionic acid
Supplier: Matrix Scientific
041574-1G
041574-5G
101852-322EA
573.85
USD
101852-322
101852-324
Fmoc-DL-2-amino-3-(dimethylamino)propionic acid
Fmoc-DL-2-amino-3-(dimethylamino)propionic acid
Hazard: Irritant
Formula:
C₂₀H₂₂N₂O₄ |
MDL Number:
MFCD04974551 CAS Number: 1219391-60-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...