Fmoc-DL-(2-thienyl)glycine

Supplier: Matrix Scientific

Synonyms: 2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(thiophen-2-yl)acetic acid

041573-1G 041573-5G
101852-318EA 586.15 USD
101852-318 101852-320
Fmoc-DL-(2-thienyl)glycine
Fmoc-DL-(2-thienyl)glycine

Hazard: Irritant

Formula: C₂₁H₁₇NO₄S
MW: 379.44 g/mol
MDL Number: MFCD08458753
CAS Number: 211682-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR