Fmoc-DL-(2-thiazoyl)glycine

Supplier: Matrix Scientific

041572-500MG 041572-1G
101852-314EA 1395.38 USD
101852-314 101852-316
Fmoc-DL-(2-thiazoyl)glycine
Fmoc-DL-(2-thiazoyl)glycine

Hazard: Irritant

Formula: C₂₀H₁₆N₂O₄S
MDL Number: MFCD09750508
CAS Number: 1219381-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR